Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-NH-PEG6-CH2COOH | 100 g | CAS 437655-96-4 | MDL MFCD26142983

Catalog No. 502352505
Encompass

Fmoc-NH-PEG6-CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 437655-96-4
- MDL: MFCD26142983
- InChIKey: HANLHKUCDRJGKX-UHFFFAOYSA-N
- Molecular Weight: 561.628
- Molecular Formula: C29H39NO10
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azatetracosan-24-oic acid
- SMILES: O=C(NCCOCCOCCOCCOCCOCCOCC(O)=O)OCC1C2=C(C3=C1C=CC=C3)C=CC=C2

Catalog No. 50-235-2505 Supplier STA Pharmaceutical Supplier No. STAAA10065100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-PEG6-CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 437655-96-4
- MDL: MFCD26142983
- InChIKey: HANLHKUCDRJGKX-UHFFFAOYSA-N
- Molecular Weight: 561.628
- Molecular Formula: C29H39NO10
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azatetracosan-24-oic acid
- SMILES: O=C(NCCOCCOCCOCCOCCOCCOCC(O)=O)OCC1C2=C(C3=C1C=CC=C3)C=CC=C2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.