Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-NIP-OH (Fmoc-3-carboxypiperidine) | 158922-07-7 | ≥97.0% | 351.40 g·mol⁻¹ | C21H21NO4 | 25 G
SDP

Catalog No. 5000208838
Encompass_Preferred

Fmoc-NIP-OH (Fmoc-3-carboxypiperidine) is an Fmoc-protected amino acid derivative used as a building block in solid-phase peptide synthesis and related peptide chemistry.

  • Used as an Fmoc-protected 3-carboxypiperidine building block for peptide synthesis.
  • Purity ≥97.0%.
  • Molecular weight 351.40 g·mol⁻¹; chemical formula C21H21NO4.
  • Available in 1 g, 5 g, 10 g, 25 g, and 50 g package sizes.
  • CAS number 158922-07-7; consult the SDS for handling and safety information.

Catalog No. 50-002-08838 Supplier Medchemexpress LLC Supplier No. 25GHYW088805
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NIP-OH (Fmoc-3-carboxypiperidine) is an Fmoc-protected amino acid derivative used as a building block in solid-phase peptide synthesis and related peptide chemistry.

  • Used as an Fmoc-protected 3-carboxypiperidine building block for peptide synthesis.
  • Purity ≥97.0%.
  • Molecular weight 351.40 g·mol⁻¹; chemical formula C21H21NO4.
  • Available in 1 g, 5 g, 10 g, 25 g, and 50 g package sizes.
  • CAS number 158922-07-7; consult the SDS for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.