Learn More
MilliporeSigma FMOC-NLE-OH 5G
Catalog No. 502301647
Encompass
Fmoc-Nle-OH; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-norleucine; F.W. 353.4; C21H23NO4; M.P. 135-145 deg.C (275-293 deg.F); CAS: 772
Catalog No. 50-230-1647 Supplier MilliporeSigma Supplier No. 8520140005
Fmoc-Nle-OH; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-norleucine; F.W. 353.4; C21H23NO4; M.P. 135-145 deg.C (275-293 deg.F); CAS: 772