Learn More
Medchemexpress LLC Fmoc-NMe-PEG4-C2-acid | 2170240-98-7 | 97.0% | C27H35NO8 | 25 MG
SDP
Catalog No. 5000352579
Encompass_Preferred
Fmoc-NMe-PEG4-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-003-52579 Supplier Medchemexpress LLC Supplier No. 25MGHY140465
Add to Cart
Add to Cart
Fmoc-NMe-PEG4-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs