Learn More
Medchemexpress LLC Fmoc-NMe-Val-Val-Dil-Dap-OH | 1352202-13-1 | 96.2% | 807.03 | C45H66N4O9 | 500 MG
SDP
Catalog No. 5000281396
Encompass_Preferred
Fmoc-NMe-Val-Val-Dil-Dap-OH is an Fmoc-protected peptide intermediate used to build drug-linker conjugates for antibody-drug conjugate (ADC) research. Supplied as a solid powder for research use, the compound is characterized by molecular formula C45H66N4O9, molecular weight 807.03, and a reported supplier purity of ~96.2%.
  • Fmoc-protected peptide intermediate suitable for linker synthesis.
  • Intended for preparation of drug-linker conjugates used in ADC construction.
  • Supplied as a solid powder for laboratory handling and storage.
  • Molecular weight 807.03 and formula C45H66N4O9 to support formulation planning.
  • Reported high purity suitable for research applications.
Catalog No. 50-002-81396 Supplier Medchemexpress LLC Supplier No. HYW250408500MG
Fmoc-NMe-Val-Val-Dil-Dap-OH is an Fmoc-protected peptide intermediate used to build drug-linker conjugates for antibody-drug conjugate (ADC) research. Supplied as a solid powder for research use, the compound is characterized by molecular formula C45H66N4O9, molecular weight 807.03, and a reported supplier purity of ~96.2%.
  • Fmoc-protected peptide intermediate suitable for linker synthesis.
  • Intended for preparation of drug-linker conjugates used in ADC construction.
  • Supplied as a solid powder for laboratory handling and storage.
  • Molecular weight 807.03 and formula C45H66N4O9 to support formulation planning.
  • Reported high purity suitable for research applications.