Learn More
Medchemexpress LLC Fmoc-OSu | 82911-69-1 | >99.8% | C19H15NO5 | 25 G
SDP
Catalog No. 5000357965
Encompass_Preferred
Fmoc-OSu (N-(Fmoc-oxy)succinimide) is an acylating agent that selectively targets amino groups, protecting them during peptide synthesis by forming stable amide bonds. It also serves as a fluorescent labeling reagent, notably for N-sugar chains, and is applied in N-glycan labeling experiments.
  • Selectively protects amino groups
  • Forms stable amide bonds for peptide synthesis
  • Functions as a fluorescent labeling reagent
  • Used in N-glycan labeling experiments
Catalog No. 50-003-57965 Supplier Medchemexpress LLC Supplier No. 25GHY79647
Fmoc-OSu (N-(Fmoc-oxy)succinimide) is an acylating agent that selectively targets amino groups, protecting them during peptide synthesis by forming stable amide bonds. It also serves as a fluorescent labeling reagent, notably for N-sugar chains, and is applied in N-glycan labeling experiments.
  • Selectively protects amino groups
  • Forms stable amide bonds for peptide synthesis
  • Functions as a fluorescent labeling reagent
  • Used in N-glycan labeling experiments