Learn More
Medchemexpress LLC Fmoc-PEG1-CH2CH2-NHS ester | 1807521-05-6 | 96.3% | 250 MG
SDP
Catalog No. 5000341985
Encompass_Preferred
Fmoc-PEG1-CH2CH2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteeasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-41985 Supplier Medchemexpress LLC Supplier No. 250MGHY130077
Fmoc-PEG1-CH2CH2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteeasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins