Learn More
Medchemexpress LLC Fmoc-PEG3-alcohol | 560088-66-6 | >98.0% | 371.43 g/mol | C21H25NO5 | 500 MG
SDP
Catalog No. 5000283025
Encompass_Preferred
Fmoc-PEG3-alcohol is an Fmoc-protected PEG3 spacer terminating in a primary alcohol. Supplied as a white to off-white solid, it is used as a short, flexible linker in the synthesis of PROTACs, bifunctional molecules, and medicinal chemistry intermediates where an easily removable protecting group and a derivatizable hydroxy terminus are required.
  • Fmoc-protected amine enables orthogonal protection and deprotection workflows.
  • PEG3 spacer adds flexibility and improves aqueous solubility.
  • Terminal primary alcohol allows for esterification, etherification, or activation for coupling.
  • Solid form is easy to weigh and handle in laboratory synthesis.
  • Suitable for PROTAC synthesis, bioconjugation, and linker optimization studies.
  • Offered in multiple lab-scale packaging sizes for convenience.
Catalog No. 50-002-83025 Supplier Medchemexpress LLC Supplier No. HYW143484500MG
Fmoc-PEG3-alcohol is an Fmoc-protected PEG3 spacer terminating in a primary alcohol. Supplied as a white to off-white solid, it is used as a short, flexible linker in the synthesis of PROTACs, bifunctional molecules, and medicinal chemistry intermediates where an easily removable protecting group and a derivatizable hydroxy terminus are required.
  • Fmoc-protected amine enables orthogonal protection and deprotection workflows.
  • PEG3 spacer adds flexibility and improves aqueous solubility.
  • Terminal primary alcohol allows for esterification, etherification, or activation for coupling.
  • Solid form is easy to weigh and handle in laboratory synthesis.
  • Suitable for PROTAC synthesis, bioconjugation, and linker optimization studies.
  • Offered in multiple lab-scale packaging sizes for convenience.