Learn More
Medchemexpress LLC Fmoc-PEG3-alcohol | 560088-66-6 | MFCD08669697 | 95.0% | 371.43 g·mol-1 | C21H25NO5 | 100 MG
SDP
Catalog No. 5000283026
Encompass_Preferred
Fmoc-PEG3-alcohol is an Fmoc-protected polyethylene glycol (PEG) linker terminated with a primary alcohol. It functions as a short, flexible spacer for constructing bifunctional molecules and is commonly used in PROTAC synthesis and medicinal chemistry workflows.
  • Fmoc-protected carbamate enables orthogonal deprotection strategies.
  • Three-unit PEG spacer offers flexibility and improved solubility.
  • Primary alcohol terminus allows for oxidation or coupling reactions.
  • Well suited for assembling bifunctional degraders and linker construction.
  • Provided as a research-grade solid for synthetic applications.
Catalog No. 50-002-83026 Supplier Medchemexpress LLC Supplier No. HYW143484100MG
Fmoc-PEG3-alcohol is an Fmoc-protected polyethylene glycol (PEG) linker terminated with a primary alcohol. It functions as a short, flexible spacer for constructing bifunctional molecules and is commonly used in PROTAC synthesis and medicinal chemistry workflows.
  • Fmoc-protected carbamate enables orthogonal deprotection strategies.
  • Three-unit PEG spacer offers flexibility and improved solubility.
  • Primary alcohol terminus allows for oxidation or coupling reactions.
  • Well suited for assembling bifunctional degraders and linker construction.
  • Provided as a research-grade solid for synthetic applications.