Learn More
Medchemexpress LLC Fmoc-PEG4-NHS ester | 1314378-14-7 | 98.6% | 1 G
SDP
Catalog No. 5000231477
Encompass_Preferred
Fmoc-PEG4-NHS ester is a PEG-based linker that combines an Fmoc-protected amine with an N-hydroxysuccinimide (NHS) activated ester on a four-unit PEG spacer, enabling convenient conjugation to primary amines for bioconjugation and PROTAC synthesis.
  • Suitable for synthesis of PROTACs and other bioconjugates.
  • Contains an Fmoc-protected amine for orthogonal deprotection.
  • Features an NHS-activated ester for rapid amide bond formation with primary amines.
  • PEG4 spacer increases aqueous solubility and provides flexible separation between linked moieties.
  • Available in small-scale quantities with high reported purity for synthetic use.
Catalog No. 50-002-31477 Supplier Medchemexpress LLC Supplier No. HY1224561G
Fmoc-PEG4-NHS ester is a PEG-based linker that combines an Fmoc-protected amine with an N-hydroxysuccinimide (NHS) activated ester on a four-unit PEG spacer, enabling convenient conjugation to primary amines for bioconjugation and PROTAC synthesis.
  • Suitable for synthesis of PROTACs and other bioconjugates.
  • Contains an Fmoc-protected amine for orthogonal deprotection.
  • Features an NHS-activated ester for rapid amide bond formation with primary amines.
  • PEG4 spacer increases aqueous solubility and provides flexible separation between linked moieties.
  • Available in small-scale quantities with high reported purity for synthetic use.