Learn More
Medchemexpress LLC HY-114430 50mg Medchemexpress, Fmoc-Phe-Lys(Boc)-PAB-PNP CAS:1646299-50-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11443050MG
Encompass_Preferred
Medchemexpress, HY-114430 50mg Fmoc-Phe-Lys(Boc)-PAB-PNP CAS:1646299-50-4 Fmoc-Phe-Lys(Boc)-PAB-PNP is an ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2491