Learn More
Medchemexpress LLC Fmoc-Phe-OH-13C | 916262-84-5 | >99.8% | C2313CH21NO4 | 100 MG
SDP
Catalog No. 5000435046
Encompass_Preferred
Fmoc-Phe-OH-13C is a 13C labeled phenylalanine derivative used in laboratory research. It is stable under recommended storage conditions and has a high purity.
  • 13C labeled phenylalanine derivative
  • Suitable for laboratory research
  • High purity of >99.8% by HPLC
  • Stable under recommended storage conditions
  • Specific storage conditions for powder and in solvent to ensure longevity.
Catalog No. 50-004-35046 Supplier Medchemexpress LLC Supplier No. HY79131S5100MG
Fmoc-Phe-OH-13C is a 13C labeled phenylalanine derivative used in laboratory research. It is stable under recommended storage conditions and has a high purity.
  • 13C labeled phenylalanine derivative
  • Suitable for laboratory research
  • High purity of >99.8% by HPLC
  • Stable under recommended storage conditions
  • Specific storage conditions for powder and in solvent to ensure longevity.