Learn More
Medchemexpress LLC Fmoc-Phe-OH-13C9,15N | 1217455-27-0 | MFCD08702953 | 99.3% | 397.36 g·mol⁻1 | C15H21NO4 (13C9, 15N) | 10 MG
SDP
Catalog No. 5000298680
Encompass_Preferred
Isotopically labeled Fmoc-protected phenylalanine (13C9, 15N) intended for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. Supplied in small, research-scale quantities with high reported purity (99.33%).
  • Isotopically labeled with 13C9 and 15N for mass and NMR differentiation.
  • Fmoc-protected phenylalanine derivative suitable for peptide synthesis or analytical standards.
  • High purity (≈99.3%) for quantitative applications.
  • Suitable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Available in multiple small package sizes for research use.
Catalog No. 50-002-98680 Supplier Medchemexpress LLC Supplier No. HY79131S110MG
Isotopically labeled Fmoc-protected phenylalanine (13C9, 15N) intended for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. Supplied in small, research-scale quantities with high reported purity (99.33%).
  • Isotopically labeled with 13C9 and 15N for mass and NMR differentiation.
  • Fmoc-protected phenylalanine derivative suitable for peptide synthesis or analytical standards.
  • High purity (≈99.3%) for quantitative applications.
  • Suitable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Available in multiple small package sizes for research use.