Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Phe-OSu, Advanced ChemTech

Catalog No. 01340485
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-485 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-485 Supplier Advanced Chem Tech Supplier No. FF24265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 101214-43-1 | C28H24N2O6 | 484.51 g/mol

Chemical Identifiers

CAS 101214-43-1
Molecular Formula C28H24N2O6
Molecular Weight (g/mol) 484.51
MDL Number MFCD00062204
InChI Key VLXHZQQUTCVLGU-DEOSSOPVSA-N
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoate
SMILES O=C(N[C@@H](CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White to Off-White
Packaging Bottle
Quantity 5 g
Percent Purity 98%
Grade Biochemical
Chemical Name or Material Fmoc-Phe-OSu
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.