Learn More
Bachem Fmoc-Phe-Phe-OH
SDP
Supplier:  Bachem 4015688.0001
Encompass_Preferred
Fmoc-Phe-Phe-OH, 84889-09-8, C33H30N2O5, Mr 534.61, 1g. The nanotube formation of Fmoc-FF has been studied by Gazit and Reches. Jayawarna et al. found that this self-assembling dipeptide forms fibrous hydrogels at pH<8. Such hydrogels are excellently suited for 3D cell microculture formation, they can be modified by addition of Fmoc-amino acids as Fmoc-Lys-OH.
Catalog No. 50-258-618