Learn More
MilliporeSigma FMOC-PHG-OH
Catalog No. 502301811
Encompass
Fmoc-Phg-OH; > /=98% by TLC and HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-phenylglycine; F.W. 373.4; C23H19NO4; CAS: 102410-65-1
Catalog No. 50-230-1811 Supplier MilliporeSigma Supplier No. 8522130005
Fmoc-Phg-OH; > /=98% by TLC and HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-L-phenylglycine; F.W. 373.4; C23H19NO4; CAS: 102410-65-1