Learn More
Sigma Aldrich Fine Chemicals Biosciences fmoc Pro NovaSynR TGT Nova
Catalog No. 502900609
Encompass
An excellent resin for the fmoc continuous flow synthesis of peptides containing C-terminal Pro without risk of diketopiperazine formation.The product number for this product was previously 04-12-2719.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-0609 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8560490005
An excellent resin for the fmoc continuous flow synthesis of peptides containing C-terminal Pro without risk of diketopiperazine formation.The product number for this product was previously 04-12-2719.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.