Learn More
Medchemexpress LLC Fmoc-Pro-OH-13C5,15N | 1217452-48-6 | 343.33 | 5 MG
SDP
Catalog No. 5000454587
Encompass_Preferred
Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled proline derivative, specifically an Fmoc-Pro-OH compound. It incorporates stable heavy isotopes of carbon and nitrogen, making it suitable for use as a tracer in drug development for quantitation and studying the impact of isotopic substitution on drug pharmacokinetics and metabolic profiles.
  • 15N-labeled and 13C-labeled proline derivative
  • Useful as a tracer in drug development
  • Aids in quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research purposes only
Catalog No. 50-004-54587 Supplier Medchemexpress LLC Supplier No. HYW013780S15MG
Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled proline derivative, specifically an Fmoc-Pro-OH compound. It incorporates stable heavy isotopes of carbon and nitrogen, making it suitable for use as a tracer in drug development for quantitation and studying the impact of isotopic substitution on drug pharmacokinetics and metabolic profiles.
  • 15N-labeled and 13C-labeled proline derivative
  • Useful as a tracer in drug development
  • Aids in quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research purposes only