Learn More
Medchemexpress LLC Fmoc-Pro-OH-13C5,15N | 1217452-48-6 | MFCD08702954 | 98.0% | 343.33 | 100 MG
SDP
Catalog No. 5000454635
Encompass_Preferred
Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Pro-OH, a proline derivative. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labeled compound.
  • Proline derivative.
  • Used as tracers for quantitation during drug development.
  • Relevant for studies on pharmacokinetic and metabolic profiles of drugs.
  • Targeted for fungal and isotope-labeled compounds.
  • Applicable in anti-infection and other pathways.
Catalog No. 50-004-54635 Supplier Medchemexpress LLC Supplier No. HYW013780S1100MG
Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Pro-OH, a proline derivative. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labeled compound.
  • Proline derivative.
  • Used as tracers for quantitation during drug development.
  • Relevant for studies on pharmacokinetic and metabolic profiles of drugs.
  • Targeted for fungal and isotope-labeled compounds.
  • Applicable in anti-infection and other pathways.