Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Pro-OH-13C5,15N | 1217452-48-6 | MFCD08702954 | 98% | 343.33 | 10 MG
SDP

Catalog No. 5000454449
Encompass_Preferred

Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled proline derivative (Fmoc-Pro-OH). Stable heavy isotopes like these are incorporated into drug molecules for quantitation during drug development processes. This compound is also suitable for peptide synthesis.

  • Isotopic purity 98 atom % 13C, 98 atom % 15N
  • Solid form
  • Suitable for bio NMR technique
  • Melting point 117-118 °C
  • Fmoc functional group
  • Mass shift M+6
  • Recommended storage at -20°C

Catalog No. 50-004-54449 Supplier Medchemexpress LLC Supplier No. HYW013780S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Pro-OH-13C5,15N is a 15N-labeled and 13C-labeled proline derivative (Fmoc-Pro-OH). Stable heavy isotopes like these are incorporated into drug molecules for quantitation during drug development processes. This compound is also suitable for peptide synthesis.

  • Isotopic purity 98 atom % 13C, 98 atom % 15N
  • Solid form
  • Suitable for bio NMR technique
  • Melting point 117-118 °C
  • Fmoc functional group
  • Mass shift M+6
  • Recommended storage at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.