Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Pro-OH, Advanced ChemTech

Catalog No. 01340117
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-117 25 g
01-340-119 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-340-117 Supplier Advanced Chem Tech Supplier No. FP245025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71989-31-6
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.38
InChI Key ZPGDWQNBZYOZTI-UHFFFAOYNA-N
IUPAC Name 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-Pro-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.