Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-pro-pro-oh | 129223-22-9 | MFCD00237673 | 99.8% | 434.48 g/mol | C25H26N2O5 | 10 G
SDP

Catalog No. 5000209249
Encompass_Preferred

Fmoc-Pro-Pro-OH is an Fmoc-protected proline dipeptide used as a building block in solid-phase peptide synthesis and related peptide chemistry. It is supplied as a white to off-white solid for research use only.

  • High purity: 99.8% (as supplied)
  • Physical form: white to off-white solid powder
  • Chemical formula: C25H26N2O5
  • Molecular weight: 434.48 g/mol
  • Solubility: soluble in DMSO (≈66.7 mg/mL with warming and sonication)
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solution keep at -80°C or -20°C for limited times
  • Intended use: reagent for peptide synthesis and laboratory research

Catalog No. 50-002-09249 Supplier Medchemexpress LLC Supplier No. HYW04199010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Pro-Pro-OH is an Fmoc-protected proline dipeptide used as a building block in solid-phase peptide synthesis and related peptide chemistry. It is supplied as a white to off-white solid for research use only.

  • High purity: 99.8% (as supplied)
  • Physical form: white to off-white solid powder
  • Chemical formula: C25H26N2O5
  • Molecular weight: 434.48 g/mol
  • Solubility: soluble in DMSO (≈66.7 mg/mL with warming and sonication)
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solution keep at -80°C or -20°C for limited times
  • Intended use: reagent for peptide synthesis and laboratory research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.