Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-pro-pro-oh | 129223-22-9 | MFCD00237673 | 99.8% | 434.48 g/mol | C25H26N2O5 | 25 G
SDP

Catalog No. 5000209318
Encompass_Preferred

Fmoc-protected dipeptide of two proline residues supplied as a white to off-white solid for use as a building block in peptide synthesis and related research applications. For research use only.

  • High purity suitable for synthesis and analytical applications.
  • White to off-white solid with defined storage recommendations for powder and solutions.
  • Molecular weight 434.48 g/mol and chemical formula C25H26N2O5.
  • Certificate of analysis available confirming reported purity.
  • Available in multiple pack sizes, including 25 G, to support small-scale synthesis.

Catalog No. 50-002-09318 Supplier Medchemexpress LLC Supplier No. 25GHYW041990
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-protected dipeptide of two proline residues supplied as a white to off-white solid for use as a building block in peptide synthesis and related research applications. For research use only.

  • High purity suitable for synthesis and analytical applications.
  • White to off-white solid with defined storage recommendations for powder and solutions.
  • Molecular weight 434.48 g/mol and chemical formula C25H26N2O5.
  • Certificate of analysis available confirming reported purity.
  • Available in multiple pack sizes, including 25 G, to support small-scale synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.