Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-(R)-2-(7-octenyl)Ala-OH | 945212-26-0 | 421.53 | 250 MG
SDP

Catalog No. 5000449957
Encompass_Preferred

Fmoc-(R)-2-(7-octenyl)Ala-OH is an amino acid derivative with an Fmoc protecting group. This compound can be utilized to synthesize inhibitor peptides that combinatorially inactivate ErbB1, ErbB2, and ErbB3.

  • Chemical formula: C26H31NO4
  • Appearance: Solid-liquid mixture
  • Color: Off-white to yellow
  • Storage: 4°C, stored under nitrogen (solid); -80°C for 6 months, -20°C for 1 month in solvent (stored under nitrogen)
  • Solubility: 200 mg/mL in DMSO

Catalog No. 50-004-49957 Supplier Medchemexpress LLC Supplier No. 250MGHYW006886
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-(R)-2-(7-octenyl)Ala-OH is an amino acid derivative with an Fmoc protecting group. This compound can be utilized to synthesize inhibitor peptides that combinatorially inactivate ErbB1, ErbB2, and ErbB3.

  • Chemical formula: C26H31NO4
  • Appearance: Solid-liquid mixture
  • Color: Off-white to yellow
  • Storage: 4°C, stored under nitrogen (solid); -80°C for 6 months, -20°C for 1 month in solvent (stored under nitrogen)
  • Solubility: 200 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.