Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-(S)-2-(4-pentenyl)Ala-OH | 288617-73-2 | 379.45 | 5 G
SDP

Catalog No. 5000446063
Encompass_Preferred

Fmoc-(S)-2-(4-pentenyl)Ala-OH is an Fmoc-protected unnatural amino acid used in polypeptide synthesis.

  • Fmoc-protected unnatural amino acid
  • Suitable for polypeptide synthesis
  • Appears as a solid
  • Color is white to light yellow
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-46063 Supplier Medchemexpress LLC Supplier No. HYW0048645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-(S)-2-(4-pentenyl)Ala-OH is an Fmoc-protected unnatural amino acid used in polypeptide synthesis.

  • Fmoc-protected unnatural amino acid
  • Suitable for polypeptide synthesis
  • Appears as a solid
  • Color is white to light yellow
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.