Learn More
Medchemexpress LLC Fmoc-(S)-2-(4-pentenyl)Ala-OH | 288617-73-2 | 379.45 | 500 MG
SDP
Catalog No. 5000449089
Encompass_Preferred
Fmoc-(S)-2-(4-pentenyl)Ala-OH is an Fmoc-protected unnatural amino acid primarily used for polypeptide synthesis, including compounds like NYAD-13. It is a key reagent for peptide synthesis, particularly in solid-phase methods.
  • Can be used for polypeptide synthesis
  • Fmoc-protected unnatural amino acid
  • Involved in protein-protein interactions
  • Used in all-hydrocarbon peptide stapling
  • Reagent for peptide synthesis
Catalog No. 50-004-49089 Supplier Medchemexpress LLC Supplier No. HYW004864500MG
Fmoc-(S)-2-(4-pentenyl)Ala-OH is an Fmoc-protected unnatural amino acid primarily used for polypeptide synthesis, including compounds like NYAD-13. It is a key reagent for peptide synthesis, particularly in solid-phase methods.
  • Can be used for polypeptide synthesis
  • Fmoc-protected unnatural amino acid
  • Involved in protein-protein interactions
  • Used in all-hydrocarbon peptide stapling
  • Reagent for peptide synthesis