Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-(S)-3-amino-3-(2-bromo-phenyl)-propionic acid

Catalog No. CDS004998 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004998 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004998 Supplier Sigma Aldrich Supplier No. CDS004998100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-dimercapto-1-propanol tributyrate is a thioester analog of triacylglycerol and has been used:
  • In lipase inhibition assay during in vitro screening of medicinal plants as antidiabetics with glucosidase and lipase inhibitory activities
  • In colorimetric microplate assay for lipase activity
  • To analyze positional specificity of lipg (lipase-encoding gene) toward triacylglycerol by a simple continuous spectrophotometric method

Specifications

Molecular Formula C24H20BrNO4
Linear Formula C24H20BrNO4
MDL Number MFCD03427999
Quantity 1 g
Molecular Weight (g/mol) 466.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.