Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-Sarcosine hydrate | 100 g | CAS 212651-47-3 | MDL MFCD00180724

Catalog No. 502343406
Encompass

Fmoc-Sarcosine hydrate is a Amino Acid reagent (Subcategory: N-Me AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 212651-47-3
- MDL: MFCD00180724
- InChIKey: CUJSWOOWOONPRH-UHFFFAOYSA-N
- Molecular Weight: 329.352
- Molecular Formula: C18H19NO5
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-methylglycine hydrate
- SMILES: CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)CC(O)=O.O

Catalog No. 50-234-3406 Supplier STA Pharmaceutical Supplier No. STAAA11720100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Sarcosine hydrate is a Amino Acid reagent (Subcategory: N-Me AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 212651-47-3
- MDL: MFCD00180724
- InChIKey: CUJSWOOWOONPRH-UHFFFAOYSA-N
- Molecular Weight: 329.352
- Molecular Formula: C18H19NO5
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-methylglycine hydrate
- SMILES: CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)CC(O)=O.O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.