Learn More
Sigma Aldrich Fine Chemicals Biosciences fmoc SDMABoc2 ONa Novabioc
Catalog No. 502900564
Encompass
A derivative for the introduction of symmetric dimethyl-arginine during fmoc SPPS. Coupling can be carried out using any standard activation method. Removal of the Boc protecting groups occurs during the course of the TfA-mediated cleavage reaction. Ref [1] contains methods and protocols for the synthesis of arrays of histone-related peptides containing methylated arginine and lysine-residues.
Catalog No. 50-290-0564 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8523100001
A derivative for the introduction of symmetric dimethyl-arginine during fmoc SPPS. Coupling can be carried out using any standard activation method. Removal of the Boc protecting groups occurs during the course of the TfA-mediated cleavage reaction. Ref [1] contains methods and protocols for the synthesis of arrays of histone-related peptides containing methylated arginine and lysine-residues.