Learn More
Medchemexpress LLC Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH | 120173-57-1 | MFCD00171375 | 99.0% | 656.63 g/mol | C32H36N2O13 | 50 MG
SDP
Catalog No. 5000244170
Encompass_Preferred
Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is an Fmoc-protected serine derivative bearing an O-linked α-D-N-acetylgalactosamine tris-acetate. It is supplied as a high-purity solid for research use and is intended as a synthetic building block for glycopeptide synthesis and vaccine-related constructs in laboratory workflows.
  • High purity (99.0%) for reliable synthesis results.
  • Suitable for glycopeptide and vaccine construct synthesis.
  • Available in multiple small-scale quantities for method development.
  • Solid form simplifies handling and storage in dry conditions.
  • Molecular weight: 656.63 g/mol; formula: C32H36N2O13.
Catalog No. 50-002-44170 Supplier Medchemexpress LLC Supplier No. HY10400450MG
Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is an Fmoc-protected serine derivative bearing an O-linked α-D-N-acetylgalactosamine tris-acetate. It is supplied as a high-purity solid for research use and is intended as a synthetic building block for glycopeptide synthesis and vaccine-related constructs in laboratory workflows.
  • High purity (99.0%) for reliable synthesis results.
  • Suitable for glycopeptide and vaccine construct synthesis.
  • Available in multiple small-scale quantities for method development.
  • Solid form simplifies handling and storage in dry conditions.
  • Molecular weight: 656.63 g/mol; formula: C32H36N2O13.