Learn More
Medchemexpress LLC Fmoc-ser-oh-d3 | 1380308-48-4 | 330.35 | 1 MG
SDP
Catalog No. 5000446444
Encompass_Preferred
Fmoc-Ser-OH-d3 is a deuterium labeled Fmoc-Ser-OH, intended for research use only. This compound serves as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-46444 Supplier Medchemexpress LLC Supplier No. HYW007720S1MG
Fmoc-Ser-OH-d3 is a deuterium labeled Fmoc-Ser-OH, intended for research use only. This compound serves as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS