Learn More
Medchemexpress LLC Fmoc-Ser-OH-d3 | 1380308-48-4 | 330.35 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW007720S5MG
Encompass_Preferred
Fmoc-Ser-OH-d3 is the deuterium labeled Fmoc-Ser-OH. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-46445