Learn More
Bachem Fmoc-Ser(tBu)-OH
SDP
Catalog No. 50258517
Encompass_Preferred
Fmoc-Ser(tBu)-OH, 71989-33-8, C22H25NO5, Mr 383.44, 25g. To minimize the concomitant racemization, couplings of Fmoc-Ser(OtBu)-OH with uronium/aminium reagents should be performed in the presence of collidine in place of DIEA.
Catalog No. 50-258-517 Supplier Bachem Supplier No. 4003086.0025
Add to Cart
Add to Cart
Fmoc-Ser(tBu)-OH, 71989-33-8, C22H25NO5, Mr 383.44, 25g. To minimize the concomitant racemization, couplings of Fmoc-Ser(OtBu)-OH with uronium/aminium reagents should be performed in the presence of collidine in place of DIEA.