Learn More
Bachem Fmoc-Ser(tBu)-Wang resin (100-200 mesh, 0.3-0.6 mmol/g)
SDP
Supplier:  Bachem 4028572.0005
Encompass_Preferred
Fmoc-Ser(tBu)-Wang resin (100-200 mesh, 0.3-0.6 mmol/g) , Fmoc-Ser(tBu)-4-alkoxybenzyl alcohol resin, 5g. Fmoc-Ser(tBu)-p-alkoxybenzyl alcohol resin.
Catalog No. 50-258-923