Learn More
Cayman Chemical Fmoc-Succinimide, Purity: ≥98%, CAS: 82911-69-1, 25 g
SDP
Catalog No. 502955193
Encompass_Preferred
A mixed carbamate reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxyamino acids and hydroxyamino acid esters in high yields; used in the synthesis of glycopeptides
Catalog No. 50-295-5193 Supplier Cayman Chemical Supplier No. 1386125 G
A mixed carbamate reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxyamino acids and hydroxyamino acid esters in high yields; used in the synthesis of glycopeptides