Learn More
Bachem Fmoc-Tyr(SO3H)-OH
SDP
Catalog No. 501947357
Encompass_Preferred
Fmoc-Tyr(SO3H)-OH Sodium, 181952-24-9, C24H21NO8S, Mr 483.50, 1g. Fmoc-sulfotyrosine sodium salt. Fmoc-sulfotyrosine can form the monosodium (CAS-No. 106864-37-3) or the disodium salt. A mix of both can be obtained as well. The salt form of B-2765 varies from batch to batch.
Catalog No. 50-194-7357 Supplier Bachem Supplier No. 4029106.0001
Fmoc-Tyr(SO3H)-OH Sodium, 181952-24-9, C24H21NO8S, Mr 483.50, 1g. Fmoc-sulfotyrosine sodium salt. Fmoc-sulfotyrosine can form the monosodium (CAS-No. 106864-37-3) or the disodium salt. A mix of both can be obtained as well. The salt form of B-2765 varies from batch to batch.