Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Thr(tBu)-OH | 71989-35-0 | 397.46 | 250 G
SDP

Catalog No. 5000449960
Encompass_Preferred

Fmoc-Thr(tBu)-OH is a threonine derivative. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Threonine derivative
  • Influences anabolic hormone secretion
  • Supports mental performance under stress
  • Prevents exercise-induced muscle damage
  • Recognized as beneficial ergogenic dietary substances
  • Purity of 99.92%
  • White to off-white solid appearance
  • Long-term storage at room temperature (3 years)
  • Solvent storage at -80°C (2 years) or -20°C (1 year)
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-49960 Supplier Medchemexpress LLC Supplier No. 250GHYW007706
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Thr(tBu)-OH is a threonine derivative. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Threonine derivative
  • Influences anabolic hormone secretion
  • Supports mental performance under stress
  • Prevents exercise-induced muscle damage
  • Recognized as beneficial ergogenic dietary substances
  • Purity of 99.92%
  • White to off-white solid appearance
  • Long-term storage at room temperature (3 years)
  • Solvent storage at -80°C (2 years) or -20°C (1 year)
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.