Learn More
Carbosynth LLC. Fmoc-Thr(tBu)-Wang Resin (100-200 mesh) 1% DVB, 1g
SDP
Supplier:  Carbosynth LLC. RFT1300PI1G
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: 9-Fluorenylmethoxycarbonyl-O-t-Butyl-L-Threonine-p-Alkoxybenzyl Alcohol Resin, Storage Temperature: 2-10C
Catalog No. 50-168-6908