Learn More
Apexbio Technology LLC Fmoc-Tle-OH 132684-60-7 1g
SDP
Catalog No. 5000097123
Encompass_Preferred
Fmoc-Tle-OH (132684-60-7) is an N-Fmoc-protected leucine derivative commonly utilized in peptide and protein synthesis Featuring a fluorenylmethyloxycarbonyl (Fmoc) protective group it enables controlled amino acid coupling and selective Fmoc removal under mild basic conditions supporting sequential peptide chain elongation during solid-phase peptide synthesis (SPPS) Based on these properties Fmoc-Tle-OH holds research potential in drug discovery structure-activity studies and molecular recognition investigations
Catalog No. 50-000-97123 Supplier Apexbio Technology LLC Supplier No. A67151000
Fmoc-Tle-OH (132684-60-7) is an N-Fmoc-protected leucine derivative commonly utilized in peptide and protein synthesis Featuring a fluorenylmethyloxycarbonyl (Fmoc) protective group it enables controlled amino acid coupling and selective Fmoc removal under mild basic conditions supporting sequential peptide chain elongation during solid-phase peptide synthesis (SPPS) Based on these properties Fmoc-Tle-OH holds research potential in drug discovery structure-activity studies and molecular recognition investigations