Learn More
Bachem Fmoc-Trp(Boc)-OH
SDP
Supplier:  Bachem 4017674.0025
Encompass_Preferred
Fmoc-Trp(Boc)-OH, 143824-78-6, C31H30N2O6, Mr 526.59, 25g. Sold under license to US Pat. 5,300,651 and EP 0 516 070. The use of this protected tryptophan derivative in Fmoc solid-phase peptide synthesis resulted in an improved yield of the correct product by suppressing side reactions of tryptophan.
Catalog No. 50-258-607