Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Trp(Me)-OH | 1334509-86-2 | 99.7% | 440.49 g·mol⁻¹ | C27H24N2O4 | 1 G
SDP

Catalog No. 5000210434
Encompass_Preferred

Fmoc-Trp(Me)-OH is an Fmoc-protected N-methyl derivative of tryptophan intended for use in peptide and protein synthesis. The compound has a methyl substitution on the indole nitrogen and an Fmoc protecting group on the amino terminus; it is supplied as a solid with high purity for incorporation into solid-phase peptide synthesis workflows.

  • High purity: 99.7%
  • Molecular weight: 440.49 g·mol⁻¹
  • Chemical formula: C27H24N2O4
  • Physical state: solid powder
  • Storage: powder -20°C for up to 3 years, 4°C for up to 2 years
  • Applications: suitable for solid-phase peptide synthesis

Catalog No. 50-002-10434 Supplier Medchemexpress LLC Supplier No. HYW0486881G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Trp(Me)-OH is an Fmoc-protected N-methyl derivative of tryptophan intended for use in peptide and protein synthesis. The compound has a methyl substitution on the indole nitrogen and an Fmoc protecting group on the amino terminus; it is supplied as a solid with high purity for incorporation into solid-phase peptide synthesis workflows.

  • High purity: 99.7%
  • Molecular weight: 440.49 g·mol⁻¹
  • Chemical formula: C27H24N2O4
  • Physical state: solid powder
  • Storage: powder -20°C for up to 3 years, 4°C for up to 2 years
  • Applications: suitable for solid-phase peptide synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.