Learn More
Medchemexpress LLC Fmoc-Trp(Me)-OH | 1334509-86-2 | 99.7% | 440.49 g/mol | C27H24N2O4 | 500 MG
SDP
Catalog No. 5000210863
Encompass_Preferred
Fmoc-Trp(Me)-OH is the Fmoc-protected N1-methylated derivative of tryptophan used as an amino acid building block for peptide synthesis and for preparing peptides containing an N1-methylated indole residue. It is supplied as a solid and is compatible with standard Fmoc solid-phase peptide synthesis protocols.
  • Used in Fmoc solid-phase peptide synthesis (Fmoc-SPPS).
  • Provides an N1-methylated tryptophan residue for peptide modification.
  • High purity suitable for synthetic applications.
  • Soluble in DMSO; protocols available for in vitro and in vivo formulations.
  • Available in multiple package sizes, including a 500 mg solid package.
Catalog No. 50-002-10863 Supplier Medchemexpress LLC Supplier No. HYW048688500MG
Fmoc-Trp(Me)-OH is the Fmoc-protected N1-methylated derivative of tryptophan used as an amino acid building block for peptide synthesis and for preparing peptides containing an N1-methylated indole residue. It is supplied as a solid and is compatible with standard Fmoc solid-phase peptide synthesis protocols.
  • Used in Fmoc solid-phase peptide synthesis (Fmoc-SPPS).
  • Provides an N1-methylated tryptophan residue for peptide modification.
  • High purity suitable for synthetic applications.
  • Soluble in DMSO; protocols available for in vitro and in vivo formulations.
  • Available in multiple package sizes, including a 500 mg solid package.