Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Trp(Me)-OH | 1334509-86-2 | MFCD02684512 | 99.7% | 440.49 | C27H24N2O4 | 5 G
SDP

Catalog No. 5000210864
Encompass_Preferred

Fmoc-Trp(Me)-OH is an Fmoc-protected N-methylated tryptophan derivative used as a building block for solid-phase peptide synthesis and other peptide or protein chemistry applications.

  • Purity 99.66% (reported)
  • Molecular weight 440.49 g/mol
  • Chemical formula C27H24N2O4
  • CAS number 1334509-86-2
  • Soluble in DMSO 62.5 mg/mL; requires ultrasonic for dissolution
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)
  • Available as a 5 G solid pack

Catalog No. 50-002-10864 Supplier Medchemexpress LLC Supplier No. HYW0486885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Trp(Me)-OH is an Fmoc-protected N-methylated tryptophan derivative used as a building block for solid-phase peptide synthesis and other peptide or protein chemistry applications.

  • Purity 99.66% (reported)
  • Molecular weight 440.49 g/mol
  • Chemical formula C27H24N2O4
  • CAS number 1334509-86-2
  • Soluble in DMSO 62.5 mg/mL; requires ultrasonic for dissolution
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years)
  • Available as a 5 G solid pack

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.