Learn More
MilliporeSigma FMOC-TRP-OH
Catalog No. 502301677
Encompass
Fmoc-Trp-OH; > /=98% by HPLC and TLC; White to light yellowish solid powder; EMD; Novabiochem; 100g; N-alpha-Fmoc-L-tryptophan; F.W. 426.5; C26H22N2O4; M.P. 175-185 deg.C (347-365
Catalog No. 50-230-1677 Supplier MilliporeSigma Supplier No. 8522070100
Fmoc-Trp-OH; > /=98% by HPLC and TLC; White to light yellowish solid powder; EMD; Novabiochem; 100g; N-alpha-Fmoc-L-tryptophan; F.W. 426.5; C26H22N2O4; M.P. 175-185 deg.C (347-365