Learn More
Apexbio Technology LLC Fmoc-Trp(Boc)-OH 143824-78-6 100g
SDP
Catalog No. 5000061607
Encompass_Preferred
Fmoc-Trp(Boc)-OH is an amino acid derivative featuring dual protective groups (Fmoc and Boc) designed specifically for peptide synthesis and nanomaterial construction Structurally the compound contains hydrophobic and aromatic residues which promote supramolecular self-assembly processes under defined solvent conditions Due to these self-assembling properties Fmoc-Trp(Boc)-OH facilitates the formation of peptide-based nano-structures serving as platforms for controlled drug encapsulation and targeted delivery Current biomedical research utilizes these peptide nanostructures primarily for investigating drug-loading efficacy transport characteristics and sustained drug-release mechanisms with an emphasis on their potential applications in tumor therapeutics and delivery system optimization
Catalog No. 50-000-61607 Supplier Apexbio Technology LLC Supplier No. A7979100000
Fmoc-Trp(Boc)-OH is an amino acid derivative featuring dual protective groups (Fmoc and Boc) designed specifically for peptide synthesis and nanomaterial construction Structurally the compound contains hydrophobic and aromatic residues which promote supramolecular self-assembly processes under defined solvent conditions Due to these self-assembling properties Fmoc-Trp(Boc)-OH facilitates the formation of peptide-based nano-structures serving as platforms for controlled drug encapsulation and targeted delivery Current biomedical research utilizes these peptide nanostructures primarily for investigating drug-loading efficacy transport characteristics and sustained drug-release mechanisms with an emphasis on their potential applications in tumor therapeutics and delivery system optimization