Learn More
AdipoGen Fmoc-Ttds-OH
SDP
Supplier:  AdipoGen CDXF0204G001
Encompass_Preferred
Chemical. CAS 172089-14-4. Formula C29H38N2O8. MW 542.6. Synthetic. A bifunctional cross-linker for PEGylation. PEG polymers offer numerous favorable characteristics including high water solubility, high mobility in solution, lack of toxicity and immunogenicity, and ready clearance from the body. Thus, the chemical modification of biologically active compounds, such as peptides, antibody fragments, enzymes, or small molecules with polyethylene glycol chains referred to as “PEGylation”, is an effective method to tailor molecular properties to particular applications.
Catalog No. 50-256-0869