Learn More
MilliporeSigma FMOC-TYR(PO3H2)-OH
Supplier:  MilliporeSigma 8520580001
Encompass
Fmoc-Tyr(PO3H2)-OH; > /=95% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 1g; N-alpha-Fmoc-O-phospho-L-tyrosine; F.W. 483.4; C24H22NO8P; CAS:
Catalog No. 50-230-1819