Learn More
Medchemexpress LLC Fmoc-Tyr(tBu)-OH-13C9,15N | 1217442-53-9 | 469.46 | 1 MG
SDP
Catalog No. 5000446636
Encompass_Preferred
Fmoc-Tyr(tBu)-OH-13C9,15N is a 15N-labeled and 13C-labeled Fmoc-Tyr(tBu)-OH, a tyrosine derivative. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Storage conditions: -80°C for 6 months; -20°C for 1 month.
Catalog No. 50-004-46636 Supplier Medchemexpress LLC Supplier No. HYW008016S1MG
Fmoc-Tyr(tBu)-OH-13C9,15N is a 15N-labeled and 13C-labeled Fmoc-Tyr(tBu)-OH, a tyrosine derivative. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Storage conditions: -80°C for 6 months; -20°C for 1 month.