Learn More
Medchemexpress LLC Fmoc-val-ala-aminomethyl acetate | 2505045-86-1 | 97.3% | 481.54 | C26H31N3O6 | 100 MG
SDP
Catalog No. 5000204318
Encompass_Preferred
Fmoc-Val-Ala-aminomethyl acetate is an Fmoc-protected val-ala dipeptide used as a cleavable linker component for antibody-drug conjugate (ADC) synthesis in research settings. It provides an Fmoc-protected N-terminus and an aminomethyl acetate functional handle suitable for conjugation chemistry.
  • Cleavable dipeptide linker useful for ADC construction.
  • Fmoc-protected N-terminus compatible with peptide synthesis workflows.
  • Aminomethyl acetate functional handle for conjugation.
  • High purity suitable for research (97.3%).
  • Available in lab-scale quantities (25 mg-1 g).
Catalog No. 50-002-04318 Supplier Medchemexpress LLC Supplier No. HY156813100MG
Fmoc-Val-Ala-aminomethyl acetate is an Fmoc-protected val-ala dipeptide used as a cleavable linker component for antibody-drug conjugate (ADC) synthesis in research settings. It provides an Fmoc-protected N-terminus and an aminomethyl acetate functional handle suitable for conjugation chemistry.
  • Cleavable dipeptide linker useful for ADC construction.
  • Fmoc-protected N-terminus compatible with peptide synthesis workflows.
  • Aminomethyl acetate functional handle for conjugation.
  • High purity suitable for research (97.3%).
  • Available in lab-scale quantities (25 mg-1 g).