Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Val-Ala-PAB-OH | 1394238-91-5 | MFCD28557282 | 99.9% | 515.60 g/mol | C30H33N3O5 | 250 MG
SDP

Catalog No. 5000203483
Encompass_Preferred

Fmoc-Val-Ala-PAB-OH is a protease-cleavable linker used in the synthesis of antibody-drug conjugates. It combines an Fmoc-protected Val-Ala dipeptide with a para-aminobenzyl spacer and a free carboxyl group to enable coupling and self-immolative payload release. Supplied as a high-purity solid, it includes documented solubility and storage guidelines for research use.

  • Protease-cleavable Val-Ala dipeptide enables selective payload release.
  • Fmoc protection facilitates solid-phase peptide and conjugation chemistry.
  • Para-aminobenzyl spacer provides self-immolative release upon cleavage.
  • High purity (99.88%) and characterized molecular weight for reliable synthesis.
  • Soluble in DMSO with reported in vitro and in vivo formulation protocols.
  • Supplied as a solid in small-scale packages suitable for research.
  • Recommended storage at -20°C under inert atmosphere for stability.

Catalog No. 50-002-03483 Supplier Medchemexpress LLC Supplier No. HY126353250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Val-Ala-PAB-OH is a protease-cleavable linker used in the synthesis of antibody-drug conjugates. It combines an Fmoc-protected Val-Ala dipeptide with a para-aminobenzyl spacer and a free carboxyl group to enable coupling and self-immolative payload release. Supplied as a high-purity solid, it includes documented solubility and storage guidelines for research use.

  • Protease-cleavable Val-Ala dipeptide enables selective payload release.
  • Fmoc protection facilitates solid-phase peptide and conjugation chemistry.
  • Para-aminobenzyl spacer provides self-immolative release upon cleavage.
  • High purity (99.88%) and characterized molecular weight for reliable synthesis.
  • Soluble in DMSO with reported in vitro and in vivo formulation protocols.
  • Supplied as a solid in small-scale packages suitable for research.
  • Recommended storage at -20°C under inert atmosphere for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.